gRNA Design Details
- Target DNA Sequence
- ACCAAACTGTTCTGCCGCCA
- PAM Sequence
- GGG
- Target locus
-
Chr.17: 7441494 - 7441516 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
69.53/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF11
- Entrez Gene ID
-
2256
- Gene Name
- fibroblast growth factor 11
- Gene Aliases
-
FGF-11, FHF-3, FHF3
- Chromosome Location
-
Chr.17: 7438273 - 7444932 on GRCh38
- UniGene ID
- Hs.655193
- Species
-
Human