gRNA Design Details
- Target DNA Sequence
- ACGAGCAATTTCATCACAGA
- PAM Sequence
- TGG
- Target locus
-
Chr.9: 37655060 - 37655082 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.9/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tbrg1
- Entrez Gene ID
-
21376
- Gene Name
- transforming growth factor beta regulated gene 1
- Gene Aliases
-
AA408552, AA409675, Niam, Tb-5, mFLJ00213
- Chromosome Location
-
Chr.9: 37649182 - 37657312 on GRCm38
- UniGene ID
- Mm.28689
- Species
-
Mouse