gRNA Design Details
- Target DNA Sequence
- CCATGGAACCTGCCGGTTTT
- PAM Sequence
- TGG
- Target locus
-
Chr.6: 86263229 - 86263251 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
90.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tgfa
- Entrez Gene ID
-
21802
- Gene Name
- transforming growth factor alpha
- Gene Aliases
-
wa-1, wa1
- Chromosome Location
-
Chr.6: 86195251 - 86275449 on GRCm38
- UniGene ID
- Mm.137222
- Species
-
Mouse