gRNA Design Details
- Target DNA Sequence
- CTTTACCCTTCCGCCTCTTG
- PAM Sequence
- CGG
- Target locus
-
Chr.4: 57110060 - 57110082 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
61.16/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IGFBP7
- Entrez Gene ID
-
3490
- Gene Name
- insulin like growth factor binding protein 7
- Gene Aliases
-
AGM, FSTL2, IBP-7, IGFBP-7, IGFBP-7v, IGFBPRP1, MAC25, PSF, RAMSVPS, TAF
- Chromosome Location
-
Chr.4: 57031071 - 57110385 on GRCh38
- UniGene ID
- Hs.479808
- Species
-
Human