gRNA Design Details
- Target DNA Sequence
- GGCATCCGAAGAGGAGGAAG
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 77407922 - 77407944 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
76.87/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Igfbp7
- Entrez Gene ID
-
29817
- Gene Name
- insulin-like growth factor binding protein 7
- Gene Aliases
-
AGM, Fstl2, Mac25
- Chromosome Location
-
Chr.5: 77349240 - 77408045 on GRCm38
- UniGene ID
- Mm.233470
- Species
-
Mouse