gRNA Design Details
- Target DNA Sequence
- CGGCAGATCACACAAGTGTA
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 33611796 - 33611818 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.75/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ppp2cb
- Entrez Gene ID
-
19053
- Gene Name
- protein phosphatase 2 (formerly 2A), catalytic subunit, beta isoform
- Gene Aliases
-
AI115466, D8Ertd766e, PP2Ac
- Chromosome Location
-
Chr.8: 33599621 - 33619804 on GRCm38
- UniGene ID
- Mm.288765
- Species
-
Mouse