gRNA Design Details
- Target DNA Sequence
- GATCCGGACTTGACTCCCGA
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 33124673 - 33124695 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.41/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgf18
- Entrez Gene ID
-
14172
- Gene Name
- fibroblast growth factor 18
- Gene Aliases
-
D130055P09Rik, FGF-18
- Chromosome Location
-
Chr.11: 33116970 - 33147400 on GRCm38
- UniGene ID
- Mm.339812
- Species
-
Mouse