gRNA Design Details
- Target DNA Sequence
- AAGTAGTCCAAACAGCCGCA
- PAM Sequence
- TGG
- Target locus
-
Chr.16: 28398198 - 28398220 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.38/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgf12
- Entrez Gene ID
-
14167
- Gene Name
- fibroblast growth factor 12
- Gene Aliases
-
AV114868, B230343J05Rik, FGF-12, FHF-1, Fhf1
- Chromosome Location
-
Chr.16: 28157783 - 28753243 on GRCm38
- UniGene ID
- Mm.399969
- Species
-
Mouse