gRNA Design Details
- Target DNA Sequence
- AAAGTCCTCGAACTCGGAGC
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 107723254 - 107723276 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
92.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gfi1
- Entrez Gene ID
-
14581
- Gene Name
- growth factor independent 1
- Gene Aliases
-
AW495828, Gfi-1, Pal-1, Pal1
- Chromosome Location
-
Chr.5: 107716655 - 107726031 on GRCm38
- UniGene ID
- Mm.2078
- Species
-
Mouse