gRNA Design Details

Target DNA Sequence
GCGGTTCATCTTCCTGCACA
PAM Sequence
CGG
Target locus
Chr.9: 132986732 - 132986754 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 91.29/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GFI1B
Entrez Gene ID
8328
Gene Name
growth factor independent 1B transcriptional repressor
Gene Aliases
BDPLT17, ZNF163B
Chromosome Location
Chr.9: 132945545 - 132991697 on GRCh38
UniGene ID
Hs.553160
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001135031.1 NP_001128503.1 2
NM_004188.6 NP_004179.3 6
XM_006717297.3 XP_006717360.1 2
XM_011519068.2 XP_011517370.1 4
XM_011519069.2 XP_011517371.1 3
XM_011519070.2 XP_011517372.1 4
XM_017015175.1 XP_016870664.1 3
XM_017015176.1 XP_016870665.1 4