gRNA Design Details

Target DNA Sequence
GTTTCCGGTCTGAGAACCTC
PAM Sequence
TGG
Target locus
Chr.1: 15518154 - 15518176 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 89.04/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CASP9
Entrez Gene ID
842
Gene Name
caspase 9
Gene Aliases
APAF-3, APAF3, ICE-LAP6, MCH6, PPP1R56
Chromosome Location
Chr.1: 15491401 - 15524912 on GRCh38
UniGene ID
Hs.329502
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001229.4 NP_001220.2 2
NM_001278054.1 NP_001264983.1 2
NM_032996.3 NP_127463.2 2
NR_102732.1 2
XM_005246014.2 XP_005246071.1 2
XM_011542273.2 XP_011540575.1 2