gRNA Design Details
- Target DNA Sequence
- AGGCTCAGCATGTACGCCAG
- PAM Sequence
- AGG
- Target locus
-
Chr.10: 70441528 - 70441550 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
85.96/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- NODAL
- Entrez Gene ID
-
4838
- Gene Name
- nodal growth differentiation factor
- Gene Aliases
-
HTX5
- Chromosome Location
-
Chr.10: 70431936 - 70441709 on GRCh38
- UniGene ID
- Hs.370414
- Species
-
Human