gRNA Design Details

Target DNA Sequence
AGCCAGCTTGTATTTGGGCA
PAM Sequence
TGG
Target locus
Chr.7: 100688035 - 100688057 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 86.16/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GIGYF1
Entrez Gene ID
64599
Gene Name
GRB10 interacting GYF protein 1
Gene Aliases
GYF1, PERQ1
Chromosome Location
Chr.7: 100679507 - 100694282 on GRCh38
UniGene ID
Hs.414396
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_022574.4 NP_072096.2 2
XM_005250532.1 XP_005250589.1 4
XM_011516472.1 XP_011514774.1 4
XM_011516477.1 XP_011514779.1 4
XM_017012526.1 XP_016868015.1 4
XM_017012527.1 XP_016868016.1 6
XM_017012528.1 XP_016868017.1 5
XM_017012529.1 XP_016868018.1 5
XM_017012530.1 XP_016868019.1 5