gRNA Design Details

Target DNA Sequence
GCTCCAAAGACCTGCTGTTC
PAM Sequence
TGG
Target locus
Chr.14: 38255744 - 38255766 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 84.91/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CLEC14A
Entrez Gene ID
161198
Gene Name
C-type lectin domain family 14 member A
Gene Aliases
C14orf27, CEG1, EGFR-5
Chromosome Location
Chr.14: 38254000 - 38256370 on GRCh38
UniGene ID
Hs.525307
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_175060.2 NP_778230.1 1