gRNA Design Details
- Target DNA Sequence
- CAAGAACTCAGGACGGTGAA
- PAM Sequence
- TGG
- Target locus
-
Chr.19: 18386199 - 18386221 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.77/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GDF15
- Entrez Gene ID
-
9518
- Gene Name
- growth differentiation factor 15
- Gene Aliases
-
GDF-15, MIC-1, MIC1, NAG-1, PDF, PLAB, PTGFB
- Chromosome Location
-
Chr.19: 18386158 - 18389176 on GRCh38
- UniGene ID
- Hs.616962
- Species
-
Human