gRNA Design Details
- Target DNA Sequence
- GTCGTAATTCCTTTGCACAT
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 16859010 - 16859032 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.17/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Egfr
- Entrez Gene ID
-
13649
- Gene Name
- epidermal growth factor receptor
- Gene Aliases
-
9030024J15Rik, AI552599, Erbb, Errb1, Errp, Wa5, wa-2, wa2
- Chromosome Location
-
Chr.11: 16752203 - 16913907 on GRCm38
- UniGene ID
- Mm.420648
- Species
-
Mouse