gRNA Design Details

Target DNA Sequence
TCCGGAGCTGTGATCTAAGG
PAM Sequence
AGG
Target locus
Chr.12: 102419599 - 102419621 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 89.81/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
IGF1
Entrez Gene ID
3479
Gene Name
insulin like growth factor 1
Gene Aliases
IGF-I, IGFI, MGF
Chromosome Location
Chr.12: 102395867 - 102481839 on GRCh38
UniGene ID
Hs.160562
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000618.4 NP_000609.1 3
NM_001111283.2 NP_001104753.1 3
NM_001111284.1 NP_001104754.1 3
XM_017019259.1 XP_016874748.1 3
XM_017019260.1 XP_016874749.1 3
XM_017019261.1 XP_016874750.1 3
XM_017019262.1 XP_016874751.1 3
XM_017019263.1 XP_016874752.1 3