gRNA Design Details
- Target DNA Sequence
- CAACTACACCCTCGTCGTGC
- PAM Sequence
- TGG
- Target locus
-
Chr.4: 1022453 - 1022475 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.21/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGFRL1
- Entrez Gene ID
-
53834
- Gene Name
- fibroblast growth factor receptor-like 1
- Gene Aliases
-
FGFR5, FHFR
- Chromosome Location
-
Chr.4: 1011822 - 1026898 on GRCh38
- UniGene ID
- Hs.193326
- Species
-
Human