gRNA Design Details

Target DNA Sequence
CACCTTGTCCGCCATCTTTG
PAM Sequence
GGG
Target locus
Chr.4: 1022206 - 1022228 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 85.06/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FGFRL1
Entrez Gene ID
53834
Gene Name
fibroblast growth factor receptor-like 1
Gene Aliases
FGFR5, FHFR
Chromosome Location
Chr.4: 1011822 - 1026898 on GRCh38
UniGene ID
Hs.193326
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001004356.2 NP_001004356.1 3
NM_001004358.1 NP_001004358.1 3
NM_021923.3 NP_068742.2 2
XM_011513486.1 XP_011511788.1 3