gRNA Design Details

Target DNA Sequence
GCTGCACCATCTCACACAGT
PAM Sequence
TGG
Application
Gene Knockout
Score: 84.36/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
IKBKG
Entrez Gene ID
8517
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma
Gene Aliases
AMCBX1, FIP-3, FIP3, Fip3p, IKK-gamma, IKKAP1, IKKG, IMD33, IP, IP1, IP2, IPD2, NEMO, ZC2HC9
Chromosome Location
Chr.X: 154542240 - 154565046 on GRCh38
UniGene ID
Hs.43505
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001099856.4 NP_001093326.2 2
NM_001099857.2 NP_001093327.1 2
NM_001145255.2 NP_001138727.1 2
NM_001321396.1 NP_001308325.1 2
NM_001321397.1 NP_001308326.1 2
NM_003639.4 NP_003630.1 2