gRNA Design Details

Target DNA Sequence
ATGCGGATCAAACCTCACCA
PAM Sequence
AGG
Target locus
Chr.6: 43778463 - 43778485 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 87.01/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
VEGFA
Entrez Gene ID
7422
Gene Name
vascular endothelial growth factor A
Gene Aliases
MVCD1, VEGF, VPF
Chromosome Location
Chr.6: 43770209 - 43786487 on GRCh38
UniGene ID
Hs.73793
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001025366.2 NP_001020537.2 4
NM_001025367.2 NP_001020538.2 4
NM_001025368.2 NP_001020539.2 4
NM_001025369.2 NP_001020540.2 4
NM_001025370.2 NP_001020541.2 4
NM_001033756.2 NP_001028928.1 4
NM_001171622.1 NP_001165093.1 4
NM_001171623.1 NP_001165094.1 4
NM_001171624.1 NP_001165095.1 4
NM_001171625.1 NP_001165096.1 4
NM_001171626.1 NP_001165097.1 4
NM_001171627.1 NP_001165098.1 4
NM_001171628.1 NP_001165099.1 4
NM_001171629.1 NP_001165100.1 4
NM_001171630.1 NP_001165101.1 4
NM_001204384.1 NP_001191313.1 4
NM_001204385.1 NP_001191314.1 4
NM_001287044.1 NP_001273973.1 4
NM_001317010.1 NP_001303939.1 4
NM_003376.5 NP_003367.4 4