gRNA Design Details
- Target DNA Sequence
- ATGTGATGATAAGGACAGCC
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 70456352 - 70456374 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.34/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TGFA
- Entrez Gene ID
-
7039
- Gene Name
- transforming growth factor alpha
- Gene Aliases
-
TFGA
- Chromosome Location
-
Chr.2: 70447280 - 70554015 on GRCh38
- UniGene ID
- Hs.170009
- Species
-
Human