gRNA Design Details

Target DNA Sequence
AGTACGGGAAGCCCAACAAG
PAM Sequence
AGG
Target locus
Chr.19: 4475495 - 4475517 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 90.64/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
HDGFRP2
Entrez Gene ID
84717
Gene Name
hepatoma-derived growth factor-related protein 2
Gene Aliases
HDGF-2, HDGF2, HRP-2
Chromosome Location
Chr.19: 4472196 - 4502220 on GRCh38
UniGene ID
Hs.43071
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001001520.2 NP_001001520.1 3
NM_032631.3 NP_116020.1 3
XM_011528375.2 XP_011526677.1 3
XM_011528376.2 XP_011526678.1 3
XM_011528377.2 XP_011526679.1 3