gRNA Design Details

Target DNA Sequence
TCACGTTGGTCCACATCCTG
PAM Sequence
CGG
Target locus
Chr.14: 104780117 - 104780139 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 97/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
AKT1
Entrez Gene ID
207
Gene Name
AKT serine/threonine kinase 1
Gene Aliases
AKT, CWS6, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA
Chromosome Location
Chr.14: 104769349 - 104795743 on GRCh38
UniGene ID
Hs.525622
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001014431.1 NP_001014431.1 3
NM_001014432.1 NP_001014432.1 4
NM_005163.2 NP_005154.2 3
XM_005267401.1 XP_005267458.1 3
XM_017021075.1 XP_016876564.1 4
XM_017021076.1 XP_016876565.1 4
XM_017021077.1 XP_016876566.1 3
XM_017021078.1 XP_016876567.1 4