gRNA Design Details
- Target DNA Sequence
- GAGATAACGGCAGTGGAGGT
- PAM Sequence
- GGG
- Target locus
-
Chr.11: 69816393 - 69816415 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.65/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF3
- Entrez Gene ID
-
2248
- Gene Name
- fibroblast growth factor 3
- Gene Aliases
-
HBGF-3, INT2
- Chromosome Location
-
Chr.11: 69809968 - 69819424 on GRCh38
- UniGene ID
- Hs.37092
- Species
-
Human