gRNA Design Details

Target DNA Sequence
GTTTAGTCCAGTGGGCTTGG
PAM Sequence
GGG
Target locus
Chr.1: 115286678 - 115286700 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 92.13/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
NGF
Entrez Gene ID
4803
Gene Name
nerve growth factor
Gene Aliases
Beta-NGF, HSAN5, NGFB
Chromosome Location
Chr.1: 115285915 - 115338253 on GRCh38
UniGene ID
Hs.2561
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_002506.2 NP_002497.2 3
XM_006710663.3 XP_006710726.1 2
XM_011541518.2 XP_011539820.1 3