gRNA Design Details
- Target DNA Sequence
- TCGACTTCAAGACGTGGATG
- PAM Sequence
- CGG
- Target locus
-
Chr.16: 1984442 - 1984464 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
97.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GFER
- Entrez Gene ID
-
2671
- Gene Name
- growth factor, augmenter of liver regeneration
- Gene Aliases
-
ALR, ERV1, HERV1, HPO, HPO1, HPO2, HSS
- Chromosome Location
-
Chr.16: 1984149 - 1987749 on GRCh38
- UniGene ID
- Hs.27184
- Species
-
Human