gRNA Design Details
- Target DNA Sequence
- CGGGCAGTCCTCCCTAAACT
- PAM Sequence
- TGG
- Target locus
-
Chr.16: 1984754 - 1984776 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.11/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GFER
- Entrez Gene ID
-
2671
- Gene Name
- growth factor, augmenter of liver regeneration
- Gene Aliases
-
ALR, ERV1, HERV1, HPO, HPO1, HPO2, HSS
- Chromosome Location
-
Chr.16: 1984149 - 1987749 on GRCh38
- UniGene ID
- Hs.27184
- Species
-
Human