gRNA Design Details

Target DNA Sequence
CCATGTCTTGCTGCTGTTCT
PAM Sequence
GGG
Target locus
Chr.16: 1984854 - 1984876 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 83.7/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GFER
Entrez Gene ID
2671
Gene Name
growth factor, augmenter of liver regeneration
Gene Aliases
ALR, ERV1, HERV1, HPO, HPO1, HPO2, HSS
Chromosome Location
Chr.16: 1984149 - 1987749 on GRCh38
UniGene ID
Hs.27184
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_005262.2 NP_005253.3 2