gRNA Design Details
- Target DNA Sequence
- ACACACTCCAGGCCATCGTC
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 64235956 - 64235978 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
83.68/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- VEGFB
- Entrez Gene ID
-
7423
- Gene Name
- vascular endothelial growth factor B
- Gene Aliases
-
VEGFL, VRF
- Chromosome Location
-
Chr.11: 64234584 - 64239264 on GRCh38
- UniGene ID
- Hs.732095
- Species
-
Human