gRNA Design Details

Target DNA Sequence
ACACACTCCAGGCCATCGTC
PAM Sequence
AGG
Target locus
Chr.11: 64235956 - 64235978 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 83.68/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
VEGFB
Entrez Gene ID
7423
Gene Name
vascular endothelial growth factor B
Gene Aliases
VEGFL, VRF
Chromosome Location
Chr.11: 64234584 - 64239264 on GRCh38
UniGene ID
Hs.732095
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001243733.1 NP_001230662.1 3
NM_003377.4 NP_003368.1 3