gRNA Design Details

Target DNA Sequence
GGGACTCAATTGGCACACTG
PAM Sequence
GGG
Target locus
Chr.19: 49061925 - 49061947 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 90.47/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
NTF4
Entrez Gene ID
4909
Gene Name
neurotrophin 4
Gene Aliases
GLC10, GLC1O, NT-4, NT-4/5, NT-5, NT4, NT5, NTF5
Chromosome Location
Chr.19: 49058284 - 49065076 on GRCh38
UniGene ID
Hs.266902
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_006179.4 NP_006170.1 2
XM_005258962.3 XP_005259019.1 3
XM_006723232.3 XP_006723295.1 3
XM_011527008.2 XP_011525310.1 2
XM_011527009.2 XP_011525311.1 2
XM_011527010.2 XP_011525312.1 2