gRNA Design Details
- Target DNA Sequence
- TATTCCACCCCAGCTCCGAT
- PAM Sequence
- GGG
- Target locus
-
Chr.17: 49025642 - 49025664 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.72/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IGF2BP1
- Entrez Gene ID
-
10642
- Gene Name
- insulin like growth factor 2 mRNA binding protein 1
- Gene Aliases
-
CRD-BP, CRDBP, IMP-1, IMP1, VICKZ1, ZBP1
- Chromosome Location
-
Chr.17: 48997412 - 49056145 on GRCh38
- UniGene ID
- Hs.144936
- Species
-
Human