gRNA Design Details

Target DNA Sequence
CCGTGCGCATGTTGATGTCC
PAM Sequence
AGG
Target locus
Chr.17: 21300924 - 21300946 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 87.45/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
MAP2K3
Entrez Gene ID
5606
Gene Name
mitogen-activated protein kinase kinase 3
Gene Aliases
MAPKK3, MEK3, MKK3, PRKMK3, SAPKK-2, SAPKK2
Chromosome Location
Chr.17: 21284656 - 21315240 on GRCh38
UniGene ID
Hs.514012
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001316332.1 NP_001303261.1 6
NM_002756.4 NP_002747.2 5
NM_145109.2 NP_659731.1 5
XM_005256723.2 XP_005256780.1 6
XM_011523958.2 XP_011522260.1 5
XM_011523959.1 XP_011522261.1 7
XM_017024857.1 XP_016880346.1 5
XM_017024858.1 XP_016880347.1 6
XM_017024859.1 XP_016880348.1 7