gRNA Design Details

Target DNA Sequence
CCGTAGATCCTTCTTCCTCT
PAM Sequence
TGG
Target locus
Chr.17: 21298419 - 21298441 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 87.45/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
MAP2K3
Entrez Gene ID
5606
Gene Name
mitogen-activated protein kinase kinase 3
Gene Aliases
MAPKK3, MEK3, MKK3, PRKMK3, SAPKK-2, SAPKK2
Chromosome Location
Chr.17: 21284656 - 21315240 on GRCh38
UniGene ID
Hs.514012
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001316332.1 NP_001303261.1 3
NM_002756.4 NP_002747.2 2
NM_145109.2 NP_659731.1 2
XM_005256723.2 XP_005256780.1 3
XM_011523958.2 XP_011522260.1 2
XM_011523959.1 XP_011522261.1 4
XM_017024857.1 XP_016880346.1 2
XM_017024858.1 XP_016880347.1 3
XM_017024859.1 XP_016880348.1 4