gRNA Design Details
- Target DNA Sequence
- CCTGGACCTACACGACTTCC
- PAM Sequence
- AGG
- Target locus
-
Chr.12: 55743655 - 55743677 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
79.72/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GDF11
- Entrez Gene ID
-
10220
- Gene Name
- growth differentiation factor 11
- Gene Aliases
-
BMP-11, BMP11
- Chromosome Location
-
Chr.12: 55743278 - 55757194 on GRCh38
- UniGene ID
- Hs.600883
- Species
-
Human