gRNA Design Details

Target DNA Sequence
TCATCGAGGTCCCATCAGGT
PAM Sequence
GGG
Application
Gene Knock-Out
Score: 90.62/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
IKBKG
Entrez Gene ID
8517
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma
Gene Aliases
AMCBX1, FIP-3, FIP3, Fip3p, IKK-gamma, IKKAP1, IKKG, IMD33, IP, IP1, IP2, IPD2, NEMO, ZC2HC9
Chromosome Location
Chr.X: 154542240 - 154565046 on GRCh38
UniGene ID
Hs.43505
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001099856.4 NP_001093326.2 1