gRNA Design Details
- Target DNA Sequence
- CGAAGAGTACTACGCCAAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 218346986 - 218347008 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
96.45/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TGFB2
- Entrez Gene ID
-
7042
- Gene Name
- transforming growth factor beta 2
- Gene Aliases
-
LDS4, TGF-beta2
- Chromosome Location
-
Chr.1: 218345334 - 218444619 on GRCh38
- UniGene ID
- Hs.133379
- Species
-
Human