gRNA Design Details
- Target DNA Sequence
- AATTCGCTCCACTGTGTTGA
- PAM Sequence
- GGG
- Target locus
-
Chr.7: 55143339 - 55143361 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
89.21/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- EGFR
- Entrez Gene ID
-
1956
- Gene Name
- epidermal growth factor receptor
- Gene Aliases
-
ERBB, ERBB1, HER1, NISBD2, PIG61, mENA
- Chromosome Location
-
Chr.7: 55019032 - 55207338 on GRCh38
- UniGene ID
- Hs.488293
- Species
-
Human