gRNA Design Details
- Target DNA Sequence
- CACACACTGATGCACGGGCA
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 40444006 - 40444028 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
80.13/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IGFBP4
- Entrez Gene ID
-
3487
- Gene Name
- insulin like growth factor binding protein 4
- Gene Aliases
-
BP-4, HT29-IGFBP, IBP4, IGFBP-4
- Chromosome Location
-
Chr.17: 40443424 - 40457730 on GRCh38
- UniGene ID
- Hs.462998
- Species
-
Human