gRNA Design Details

Target DNA Sequence
CACACACTGATGCACGGGCA
PAM Sequence
AGG
Target locus
Chr.17: 40444006 - 40444028 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 80.13/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
IGFBP4
Entrez Gene ID
3487
Gene Name
insulin like growth factor binding protein 4
Gene Aliases
BP-4, HT29-IGFBP, IBP4, IGFBP-4
Chromosome Location
Chr.17: 40443424 - 40457730 on GRCh38
UniGene ID
Hs.462998
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001552.2 NP_001543.2 1