gRNA Design Details

Target DNA Sequence
CTGTACCGCCAGCAAGAATC
PAM Sequence
AGG
Target locus
Chr.3: 192170562 - 192170584 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 93.04/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FGF12
Entrez Gene ID
2257
Gene Name
fibroblast growth factor 12
Gene Aliases
FGF12B, FHF1
Chromosome Location
Chr.3: 192139393 - 192727599 on GRCh38
UniGene ID
Hs.390250
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_004113.5 NP_004104.3 5
NM_021032.4 NP_066360.1 4
XM_005247227.2 XP_005247284.1 4
XM_006713538.3 XP_006713601.1 4
XM_017005879.1 XP_016861368.1 4