gRNA Design Details
- Target DNA Sequence
- ACACATACGCCTCGCGACGT
- PAM Sequence
- TGG
- Target locus
-
Chr.10: 79756819 - 79756841 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.37/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgf22
- Entrez Gene ID
-
67112
- Gene Name
- fibroblast growth factor 22
- Gene Aliases
-
2210414E06Rik, FGF-22
- Chromosome Location
-
Chr.10: 79754575 - 79757572 on GRCm38
- UniGene ID
- Mm.154211
- Species
-
Mouse