gRNA Design Details
- Target DNA Sequence
- CGTTGCCCTGTTACTGTCTG
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 30799691 - 30799713 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
79.559/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PPP2CB
- Entrez Gene ID
-
5516
- Gene Name
- protein phosphatase 2 catalytic subunit beta
- Gene Aliases
-
PP2Abeta, PP2CB
- Chromosome Location
-
Chr.8: 30785610 - 30812836 on GRCh38
- UniGene ID
- Hs.491440
- Species
-
Human