gRNA Design Details
- Target DNA Sequence
- GCTGAACGACACCAAAGGCT
- PAM Sequence
- CGG
- Application
- Gene Knock-Out
Score:
89.35/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- NDP
- Entrez Gene ID
-
4693
- Gene Name
- NDP, norrin cystine knot growth factor
- Gene Aliases
-
EVR2, FEVR, ND
- Chromosome Location
-
Chr.X: 43948776 - 43973675 on GRCh38
- UniGene ID
- Hs.522615
- Species
-
Human