gRNA Design Details
- Target DNA Sequence
- TTTCTATCATCACAGCCGTC
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 6532397 - 6532419 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.46/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ANGPT2
- Entrez Gene ID
-
285
- Gene Name
- angiopoietin 2
- Gene Aliases
-
AGPT2, ANG2
- Chromosome Location
-
Chr.8: 6499651 - 6563420 on GRCh38
- UniGene ID
- Hs.583870
- Species
-
Human