gRNA Design Details

Target DNA Sequence
GAGTCACACGTCATCGACAC
PAM Sequence
CGG
Target locus
Chr.5: 180631706 - 180631728 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 91.29/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FLT4
Entrez Gene ID
2324
Gene Name
fms related tyrosine kinase 4
Gene Aliases
FLT-4, FLT41, LMPH1A, PCL, VEGFR-3, VEGFR3
Chromosome Location
Chr.5: 180601506 - 180650298 on GRCh38
UniGene ID
Hs.646917
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_002020.4 NP_002011.2 2
NM_182925.4 NP_891555.2 2
XM_011534478.2 XP_011532780.1 2
XM_017009263.1 XP_016864752.1 2
XM_017009264.1 XP_016864753.1 2
XM_017009265.1 XP_016864754.1 2
XM_017009266.1 XP_016864755.1 2
XM_017009267.1 XP_016864756.1 2
XM_017009268.1 XP_016864757.1 2