gRNA Design Details

Target DNA Sequence
AGAGATTGCCGGGGTGAACT
PAM Sequence
GGG
Target locus
Chr.12: 4445348 - 4445370 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 88.27/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FGF6
Entrez Gene ID
2251
Gene Name
fibroblast growth factor 6
Gene Aliases
HBGF-6, HST2
Chromosome Location
Chr.12: 4434142 - 4445799 on GRCh38
UniGene ID
Hs.166015
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_020996.2 NP_066276.2 1
XM_017018995.1 XP_016874484.1 1
XM_017018996.1 XP_016874485.1 1