gRNA Design Details
- Target DNA Sequence
- GTACATGGCCTTCACCCGCA
- PAM Sequence
- AGG
- Target locus
-
Chr.10: 101770508 - 101770530 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
89.01/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF8
- Entrez Gene ID
-
2253
- Gene Name
- fibroblast growth factor 8
- Gene Aliases
-
AIGF, FGF-8, HBGF-8, HH6, KAL6
- Chromosome Location
-
Chr.10: 101770130 - 101780369 on GRCh38
- UniGene ID
- Hs.57710
- Species
-
Human