gRNA Design Details
- Target DNA Sequence
- ATGAGCTTGTTGTTACTGTG
- PAM Sequence
- GGG
- Target locus
-
Chr.17: 63634747 - 63634769 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.46/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MAP3K3
- Entrez Gene ID
-
4215
- Gene Name
- mitogen-activated protein kinase kinase kinase 3
- Gene Aliases
-
MAPKKK3, MEKK3
- Chromosome Location
-
Chr.17: 63622209 - 63696310 on GRCh38
- UniGene ID
- Hs.29282
- Species
-
Human